Learn More
CPC Scientific H-gamma-Glu-Cys-gamma-Glu-Cys-gamma-Glu-Cys-gamma-Glu-Cys-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PHYT005A
Encompass
A glutathione-derived heavy metal-detoxifying peptide of higher plants consisting of 4 units of gammaGlu-Cys.ONE-LETTER SEQUENCE: (gammaE-C)4-GMOLECULAR FORMULA: MOLECULAR WEIGHT:1004.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Grill, E. et al. Science 230, 674 (1985) Rauser, WE. Plant Physiol. 109, 1141 (1995)
Catalog No. 50-211-0153