Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phytohemagglutin-Leukoagglutinin, MP Biomedicals™

Catalog No. MP215188601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215188601 1 mg
MP215188605 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215188601 Supplier MP Biomedicals, Inc Supplier No. 0215188601
Add to Cart
Add to Cart

Valuable in a wide variety of applications in vitro. Phytohemagglutin-Leukoagglutinin, MP Biomedicals is one of two molecular species of PHA. PHA-L has high mitogenic and leucoagglutinating activity, but very low erythroagglutinating activity.

PHA consists of two molecular species and two protein forms

  • Erythroagglutinin (PHA-E) has low mitogenic activity and high erythroagglutinin activity. It is not blood group specific, but agglutination can be inhibited by certain oligosaccharides.
  • Leucoagglutinin (PHA-L) has high mitogenic and leucoagglutinating activity, but very low erythroagglutinating activity.
  • PHA-P is the protein form of PHA prior to separation and purification of erythroagglutinin and leucoagglutinin.
  • PHA-M is the mucoprotein form.
  • Conjugates are prepared from the corresponding purified lectins.

Reconstitute with water to a final concentration of 25mg/mL This concentration of the lectin has been used for iontophoresis and neuron transport. Phytohemagglutin is very valuable in a wide variety of applications in vitro, , including:

  • Blood grouping and erythrocyte polyagglutination studies
  • Mitogenic stimulation of lymphocytes
  • Lymphocyte subpopulation studies
  • Fractionation of cells and other particles
  • Histochemical studies of normal and pathological conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym PHA-L, Phaseolus vulgaris agglutinin, PHA, PHA-P, Lectin from Phaseolus vulgaris

Specifications

Quantity 1 mg
Physical Form Lyophilized Powder
Recommended Storage −20°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.