Learn More
TARGETMOL CHEMICALS INC PHYTOHEMAGGLUTININ 25MG
SDP
Catalog No. 502577391
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Phytohemagglutinin (PHA-M) is a lectin isolated from the red kidney bean induces apoptosis via increasing proapoptotic protein Bax and activating caspases-3. Anti-tumor activitypurity: 98%
Catalog No. 50-257-7391 Supplier TARGETMOL CHEMICALS INC Supplier No. T1381825MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Phytohemagglutinin (PHA-M) is a lectin isolated from the red kidney bean induces apoptosis via increasing proapoptotic protein Bax and activating caspases-3. Anti-tumor activitypurity: 98%