Learn More
Selleck Chemical LLC Phytohemagglutinin-5mg
SDP
Catalog No. 502389825
Encompass_Preferred
Phytohemagglutinin (PHA, Phaseolus vulgaris agglutinin) is expressed in Pichia pastoris using native signal peptides, or the Saccharomyces alpha-factor preprosequence, to direct proteins into the secretory pathway. Phytohemagglutinin induces apoptosis in human HEp-2 carcinoma cells via increasing proapoptotic protein Bax and activating caspases-3.
Catalog No. 50-238-9825 Supplier Selleck Chemical LLC Supplier No. S69905MG
Phytohemagglutinin (PHA, Phaseolus vulgaris agglutinin) is expressed in Pichia pastoris using native signal peptides, or the Saccharomyces alpha-factor preprosequence, to direct proteins into the secretory pathway. Phytohemagglutinin induces apoptosis in human HEp-2 carcinoma cells via increasing proapoptotic protein Bax and activating caspases-3.