Learn More
Medchemexpress LLC Phytohemagglutinin (PHA-M) | 9008-97-3 | 100 MG
SDP
Catalog No. 5000420035
Encompass_Preferred
Phytohemagglutinin (PHA-M) is the primary seed lectin found in the common bean, Phaseolus vulgaris, and acts as a T-cell activator. It stimulates human mononuclear leukocytes, which leads to the induction of ChAT mRNA expression and enhanced acetylcholine (ACh) synthesis.
  • Acts as a T-cell activator.
  • Stimulates human mononuclear leukocytes.
  • Induces ChAT mRNA expression and enhanced acetylcholine (ACh) synthesis.
  • Binds to T-cells to stimulate metabolic activity and cell division.
  • Induces dose- and time-dependent toxicity in THP-1 monocytes/macrophages.
  • Stimulates peripheral blood mononuclear cells (PBMCs) to secrete high levels of inflammatory cytokines.
  • Inhibits the growth of renal cancer cells.
  • Causes cellular morphological changes and organelle dysfunction in THP-1 cells.
Catalog No. 50-004-20035 Supplier Medchemexpress LLC Supplier No. HYN7038100MG
Add to Cart
Add to Cart
Phytohemagglutinin (PHA-M) is the primary seed lectin found in the common bean, Phaseolus vulgaris, and acts as a T-cell activator. It stimulates human mononuclear leukocytes, which leads to the induction of ChAT mRNA expression and enhanced acetylcholine (ACh) synthesis.
  • Acts as a T-cell activator.
  • Stimulates human mononuclear leukocytes.
  • Induces ChAT mRNA expression and enhanced acetylcholine (ACh) synthesis.
  • Binds to T-cells to stimulate metabolic activity and cell division.
  • Induces dose- and time-dependent toxicity in THP-1 monocytes/macrophages.
  • Stimulates peripheral blood mononuclear cells (PBMCs) to secrete high levels of inflammatory cytokines.
  • Inhibits the growth of renal cancer cells.
  • Causes cellular morphological changes and organelle dysfunction in THP-1 cells.