Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Phytohemagglutinin, Phaseolus Vulgaris

Catalog No. 08791501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-791-501 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-791-501 Supplier MP Biomedicals™ Supplier No. 08791501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phytohemagglutinin, Phaseolus Vulgaris is crude Phaseolus vulgaris-P lectin (PHA-P) from the Red Kidney Bean.

  • Crude Phaseolus vulgaris-P lectin (PHA-P) from the Red Kidney Bean.
  • Phytohemagglutinin PHA agglutinates the erythrocytes of all human blood groups as well as those of rabbit, dog, cat, mouse, sheep, and guinea pig.
  • The optimal amount of PHA required for the agglutination of erythrocytes varies with the blood from different individuals.
  • Protein or Enzyme Type: Other Proteins, Enzymes Peptides
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Form Lyophilized

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.