Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phytol | 150-86-7 | 1 G
SDP

Catalog No. 5000421745
Encompass_Preferred

Phytol is an orally active diterpenoid alcohol extracted from chlorophyll. It exhibits antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial activities.

  • Orally active diterpenoid alcohol
  • Extracted from chlorophyll
  • Exhibits antioxidant properties
  • Exhibits anti-inflammatory properties
  • Exhibits anti-schistosomiasis activity
  • Exhibits antibacterial activity
  • Purity of 99.76%
  • Molecular formula C20H40O
  • Molecular weight 296.53
  • Appearance as a yellow to brown liquid

Catalog No. 50-004-21745 Supplier Medchemexpress LLC Supplier No. HYN30751G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phytol is an orally active diterpenoid alcohol extracted from chlorophyll. It exhibits antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial activities.

  • Orally active diterpenoid alcohol
  • Extracted from chlorophyll
  • Exhibits antioxidant properties
  • Exhibits anti-inflammatory properties
  • Exhibits anti-schistosomiasis activity
  • Exhibits antibacterial activity
  • Purity of 99.76%
  • Molecular formula C20H40O
  • Molecular weight 296.53
  • Appearance as a yellow to brown liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.