Learn More
Apexbio Technology LLC Phytol 150-86-7 250g
SDP
Catalog No. 5000064057
Encompass_Preferred
Phytol is a naturally occurring diterpene alcohol derived from chlorophyll metabolism serving as an activator for retinoid X receptors (RXRs) RXRs are nuclear hormone receptors implicated in transcriptional regulation and cellular differentiation through ligand-dependent signaling pathways involving vitamins and hormones Phytol activates RXRs displaying Ki values ranging from 2 3 to 67 2 M In mammals phytol is metabolized to phytanic acid a reaction regulated by peroxisome proliferator-activated receptor (PPAR ) Additionally phytol interacts with GABA A receptors exhibiting sedative and anxiolytic properties and has been investigated as an antischistosomal candidate in animal models
Catalog No. 50-000-64057 Supplier Apexbio Technology LLC Supplier No. C5616250000
Phytol is a naturally occurring diterpene alcohol derived from chlorophyll metabolism serving as an activator for retinoid X receptors (RXRs) RXRs are nuclear hormone receptors implicated in transcriptional regulation and cellular differentiation through ligand-dependent signaling pathways involving vitamins and hormones Phytol activates RXRs displaying Ki values ranging from 2 3 to 67 2 M In mammals phytol is metabolized to phytanic acid a reaction regulated by peroxisome proliferator-activated receptor (PPAR ) Additionally phytol interacts with GABA A receptors exhibiting sedative and anxiolytic properties and has been investigated as an antischistosomal candidate in animal models