Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phytol

Catalog No. 13991210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-991-210G 10 g
13-991-250G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-991-210G Supplier Sigma Aldrich Supplier No. 13991210G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Phytol is a diterpene alcohol and is widely used as a food additive and in medicinal field
  • It undergoes mitsunobu reaction with 5-fluorouridine to yield the nucleoterpenes.phytol was used to investigate the electroantennogram response of males and females of the potato tuber moth, phthorimaea operculella to various plant volatiles
  • It was used in the synthesis of pristanic acid (2,6,10,14-tetramethylpentadecanoic acid).phytol exhibits potential anti-schistosomal properties
  • It induces sedative and anxiolytic-like effects in mice

Specifications

CAS 7541-49-3
Density 0.85 g/mL (at 25°C (literature))
Boiling Point 202°C to 204°C (10 mmHg)
Molecular Formula C20H40O
Linear Formula CH3[CH(CH3)(CH2)3]3 C(CH3)=CHCH2OH
MDL Number MFCD00002919
Quantity 10 g
Synonym 3,7,11,15-Tetramethyl-2-hexadecen-1-ol
Molecular Weight (g/mol) 296.53
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.