Learn More
Medchemexpress LLC Phytol (Standard) | 150-86-7 | 100 MG
SDP
Catalog No. 5000421222
Encompass_Preferred
Phytol (Standard) is an analytical standard of Phytol, derived from chlorophyll, and is suitable for research and analytical applications. This orally active diterpenoid alcohol exhibits antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial properties.
  • Analytical standard for qualitative, quantitative, and methodological research
  • Used in HPLC, GC, and MS experiments
  • Possesses antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial activities
Catalog No. 50-004-21222 Supplier Medchemexpress LLC Supplier No. HYN3075R100MG
Phytol (Standard) is an analytical standard of Phytol, derived from chlorophyll, and is suitable for research and analytical applications. This orally active diterpenoid alcohol exhibits antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial properties.
  • Analytical standard for qualitative, quantitative, and methodological research
  • Used in HPLC, GC, and MS experiments
  • Possesses antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial activities