Learn More
Medchemexpress LLC Phytol (Standard) | 150-86-7 | 25 MG
SDP
Catalog No. 5000421815
Encompass_Preferred
Phytol (Standard) is an orally active diterpenoid alcohol that serves as an analytical standard. It can be extracted from chlorophyll and exhibits antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial activities. This product is intended for research and analytical applications.
  • Analytical standard for research and analytical applications
  • Extracted from chlorophyll
  • Exhibits antioxidant activity
  • Exhibits anti-inflammatory activity
  • Exhibits anti-schistosomiasis activity
  • Exhibits antibacterial activity
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-21815 Supplier Medchemexpress LLC Supplier No. 25MGHYN3075R
Phytol (Standard) is an orally active diterpenoid alcohol that serves as an analytical standard. It can be extracted from chlorophyll and exhibits antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial activities. This product is intended for research and analytical applications.
  • Analytical standard for research and analytical applications
  • Extracted from chlorophyll
  • Exhibits antioxidant activity
  • Exhibits anti-inflammatory activity
  • Exhibits anti-schistosomiasis activity
  • Exhibits antibacterial activity
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS