Learn More
Medchemexpress LLC Phytol (Standard) | 150-86-7 | 50 MG
SDP
Catalog No. 5000421902
Encompass_Preferred
Phytol (Standard) is an analytical standard used for research and analytical applications, particularly in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS. This orally active diterpenoid alcohol, extracted from chlorophyll, exhibits antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial activities. It is intended for research use only.
  • Analytical standard for research and analytical applications
  • Suitable for HPLC, GC, and MS research experiments
  • Orally active diterpenoid alcohol extracted from chlorophyll
  • Exhibits antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial activities
  • For research use only
Catalog No. 50-004-21902 Supplier Medchemexpress LLC Supplier No. HYN3075R50MG
Phytol (Standard) is an analytical standard used for research and analytical applications, particularly in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS. This orally active diterpenoid alcohol, extracted from chlorophyll, exhibits antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial activities. It is intended for research use only.
  • Analytical standard for research and analytical applications
  • Suitable for HPLC, GC, and MS research experiments
  • Orally active diterpenoid alcohol extracted from chlorophyll
  • Exhibits antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial activities
  • For research use only