Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indofine Chemical Phytolaccagenin, 98%, 20 Mg
SDP

Catalog No. 501645936
Encompass_Preferred

PHYTOLACCAGENIN, Herbal Standards, 1802-12-6, >95% by HPLC, 20 mg

Catalog No. 50-164-5936 Supplier Indofine Chemical Supplier No. 061014 20 MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PHYTOLACCAGENIN, Herbal Standards, 1802-12-6, >95% by HPLC, 20 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.