Learn More
U.S. Pharmacopeia Phytonadione, 84-80-0, MFCD00214063, 100 mg x 5 ampules
Catalog No. NC9911491
Encompass
Molecular formula C31H46O2, Molecular weight 450.7, Melting point -4 °F (-20 °C), Boiling point 284 - 293 °F (140 - 145 °C), Flash point > 235.4 °F (> 113.0 °C) Closed Cup, Synonyms: Vitamin K1, Phylloquinone, Phytomenadione
Catalog No. NC9911491 Supplier U.S. Pharmacopeia Supplier No. 1538006
Molecular formula C31H46O2, Molecular weight 450.7, Melting point -4 °F (-20 °C), Boiling point 284 - 293 °F (140 - 145 °C), Flash point > 235.4 °F (> 113.0 °C) Closed Cup, Synonyms: Vitamin K1, Phylloquinone, Phytomenadione