Learn More
Enzo Life Sciences Phytosphingosine (25 mg). CAS: 554-62-1
SDP
Supplier:  Enzo Life Sciences BMLSL1500025
Encompass_Preferred
An endogenous sphingosine occurring in Saccharomyces cerevisiae. Inhibits FMLP-induced oxidant release by human neutrophils. Purity: ≥98% (TLC). Long Term Storage: -20°C.
Catalog No. 50-201-2123