Learn More
Medchemexpress LLC Phytosphingosine | 554-62-1 | 99.9% | 317.51 | 100 MG
SDP
Catalog No. 5000453508
Encompass_Preferred
Phytosphingosine is a phospholipid exhibiting anti-inflammatory, antibacterial, and anti-cancer activities, capable of inducing apoptosis. It functions as an immune regulator and is utilized in studies of inflammatory skin diseases. Additionally, Phytosphingosine activates GPR120 with an IC50 value of 33.4 μM, making it relevant for type II diabetes research.
  • Anti-inflammatory activity
  • Antibacterial activity
  • Anti-cancer activities
  • Induces apoptosis
  • Immune regulator
  • Activates GPR120
  • Used in studies of inflammatory skin diseases
  • Used in studies of type II diabetes
Catalog No. 50-004-53508 Supplier Medchemexpress LLC Supplier No. HYW011303100MG
Phytosphingosine is a phospholipid exhibiting anti-inflammatory, antibacterial, and anti-cancer activities, capable of inducing apoptosis. It functions as an immune regulator and is utilized in studies of inflammatory skin diseases. Additionally, Phytosphingosine activates GPR120 with an IC50 value of 33.4 μM, making it relevant for type II diabetes research.
  • Anti-inflammatory activity
  • Antibacterial activity
  • Anti-cancer activities
  • Induces apoptosis
  • Immune regulator
  • Activates GPR120
  • Used in studies of inflammatory skin diseases
  • Used in studies of type II diabetes