Learn More
Medchemexpress LLC Phytosphingosine | 554-62-1 | 99.9% | 317.51 | 50 MG
SDP
Catalog No. 5000453646
Encompass_Preferred
Phytosphingosine (4-Hydroxysphinganine) is a phospholipid that exhibits anti-inflammatory, antibacterial, and anti-cancer properties. It functions as an immune regulator and an activator of GPR120 with an IC50 value of 33.4 μM. This compound is useful for research into inflammatory skin diseases and type II diabetes.
  • Induces apoptosis
  • Effective against inflammation
  • Possesses antibacterial activity
  • Exhibits anti-cancer activities
Catalog No. 50-004-53646 Supplier Medchemexpress LLC Supplier No. HYW01130350MG
Phytosphingosine (4-Hydroxysphinganine) is a phospholipid that exhibits anti-inflammatory, antibacterial, and anti-cancer properties. It functions as an immune regulator and an activator of GPR120 with an IC50 value of 33.4 μM. This compound is useful for research into inflammatory skin diseases and type II diabetes.
  • Induces apoptosis
  • Effective against inflammation
  • Possesses antibacterial activity
  • Exhibits anti-cancer activities