Learn More
Cayman Chemical Phytosphingosine (yeast), Purity: ≥95%, CAS: 554-62-1, 50 mg
SDP
Catalog No. 502982680
Encompass_Preferred
A sphingolipid; metabolized to odd-numbered fatty acids with 2-hydroxy palmitic acid as an intermediate; induces cell death in vitro; inhibits carbachol-induced PLD activation in CHO cells transfected with C. elegans muscarinic acetylcholine receptors; essential in the yeast heat stress response
Catalog No. 50-298-2680 Supplier Cayman Chemical Supplier No. 2021750 MG
A sphingolipid; metabolized to odd-numbered fatty acids with 2-hydroxy palmitic acid as an intermediate; induces cell death in vitro; inhibits carbachol-induced PLD activation in CHO cells transfected with C. elegans muscarinic acetylcholine receptors; essential in the yeast heat stress response