Learn More
Enzo Life Sciences PI-103 (1mg). CAS: 371935-74-9
SDP
Catalog No. 502008582
Encompass_Preferred
Potent, cell permeable and ATP-competitive inhibitor of DNA-PK, phosphoinositide 3-kinase (PI(3)K), the rapamycin-sensitive (mTORC1) and rapamycin-insensitive (mTORC2) complexes of the protein kinase mTOR. Exhibits little activity towards a panel of more than 40 other kinases even at concentrations as high as 10µM. Effectively blocks cell proliferation in glioma cell lines both in vitro and in vivo. Alternative name: 3-(4-(4-Morpholinyl)pyrido[3',2':4,5]furo[3,2-d]pyrimidin-2-yl)phenol. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (5mg/ml; warm). Long Term Storage: -20°C.
Catalog No. 50-200-8582 Supplier Enzo Life Sciences Supplier No. ALX270460M001
Potent, cell permeable and ATP-competitive inhibitor of DNA-PK, phosphoinositide 3-kinase (PI(3)K), the rapamycin-sensitive (mTORC1) and rapamycin-insensitive (mTORC2) complexes of the protein kinase mTOR. Exhibits little activity towards a panel of more than 40 other kinases even at concentrations as high as 10µM. Effectively blocks cell proliferation in glioma cell lines both in vitro and in vivo. Alternative name: 3-(4-(4-Morpholinyl)pyrido[3',2':4,5]furo[3,2-d]pyrimidin-2-yl)phenol. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (5mg/ml; warm). Long Term Storage: -20°C.