Learn More
Selleck Chemical LLC PI-103 S1038-1g
SDP
Catalog No. 503108513
Encompass_Preferred
PI-103 is a multi-targeted PI3K inhibitor for p110 / / / with IC50 of 2 nM/3 nM/3 nM/15 nM in cell-free assays less potent to mTOR/DNA-PK with IC50 of 30 nM/23 nM PI-103 induces apoptosis in murine T-cell Lymphoma
Catalog No. 50-310-8513 Supplier Selleck Chemical LLC Supplier No. S10381G
PI-103 is a multi-targeted PI3K inhibitor for p110 / / / with IC50 of 2 nM/3 nM/3 nM/15 nM in cell-free assays less potent to mTOR/DNA-PK with IC50 of 30 nM/23 nM PI-103 induces apoptosis in murine T-cell Lymphoma