Learn More
Medchemexpress LLC PI-103 | 371935-74-9 | 99.8% | 348.36 | 25 MG
SDP
Catalog No. 5000347664
Encompass_Preferred
PI-103 is a potent PI3K and mTOR inhibitor that also inhibits DNA-PK and induces autophagy. It shows antiproliferative effects in human cancer cell lines, leading to cell cycle arrest, inhibition of leukemic proliferation, and mitochondrial apoptosis. In vivo, it demonstrates therapeutic activity against human tumor xenografts, inhibiting angiogenesis, invasion, and metastasis.
  • Potent PI3K and mTOR inhibitor
  • Inhibits DNA-PK
  • Induces autophagy
  • Demonstrates antiproliferative properties in human cancer cell lines
  • Induces cell cycle arrest in G1 phase
  • Inhibits leukemic proliferation and mitochondrial apoptosis
  • Exhibits therapeutic activity against human tumor xenografts
  • Inhibits angiogenesis, invasion, and metastasis
  • Provides direct antiproliferative effects
Catalog No. 50-003-47664 Supplier Medchemexpress LLC Supplier No. 25MGHY10115
PI-103 is a potent PI3K and mTOR inhibitor that also inhibits DNA-PK and induces autophagy. It shows antiproliferative effects in human cancer cell lines, leading to cell cycle arrest, inhibition of leukemic proliferation, and mitochondrial apoptosis. In vivo, it demonstrates therapeutic activity against human tumor xenografts, inhibiting angiogenesis, invasion, and metastasis.
  • Potent PI3K and mTOR inhibitor
  • Inhibits DNA-PK
  • Induces autophagy
  • Demonstrates antiproliferative properties in human cancer cell lines
  • Induces cell cycle arrest in G1 phase
  • Inhibits leukemic proliferation and mitochondrial apoptosis
  • Exhibits therapeutic activity against human tumor xenografts
  • Inhibits angiogenesis, invasion, and metastasis
  • Provides direct antiproliferative effects