Learn More
Medchemexpress LLC HY-10115 5mg , PI-103 CAS:371935-74-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101155MG
Encompass_Preferred
HY-10115 5mg PI-103 CAS: 371935-74-9 PI-103 is a potent PI3K and mTOR inhibitor with IC50s of 8 nM, 88 nM, 48 nM, 150 nM, 20 nM, and 83 nM for p110α, p110β, p110δ, p110γ, mTORC1, and mTORC2. PI-103 also inhibits DNA-PK with an IC50 of 2 nM. PI-103 induces autophagy[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9422