Learn More
Selleck Chemical LLC PI-103 5mg 371935-74-9
SDP
Supplier:  Selleck Chemical LLC S10385MG
Encompass_Preferred
PI-103 is a multi-targeted PI3K inhibitor for p110?/?/?/? with IC50 of 2 nM/3 nM/3 nM/15 nM in cell-free assays, less potent to mTOR/DNA-PK with IC50 of 30 nM/23 nM. PI-103 induces apoptosis in murine T-cell Lymphoma. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-700-8