Learn More
Medchemexpress LLC HY-10115A 5mg Medchemexpress, PI-103 (Hydrochloride) CAS:371935-79-4 Purity:>98%
SDP
Catalog No. 501932056
Encompass_Preferred
Medchemexpress, HY-10115A 5mg PI-103 (Hydrochloride) CAS:371935-79-4 PI-103 Hydrochloride is a dual PI3K and mTOR inhibitor with IC 50 s of 8 nM, 88 nM, 48 nM, 150 nM, 20 nM, and 83 nM for p110α , p110β , p110δ , p110γ , mTORC1 , and mTORC2 . PI-103 also inhibits DNA-PK with an IC50 of 2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2056 Supplier Medchemexpress LLC Supplier No. HY10115A5MG
Medchemexpress, HY-10115A 5mg PI-103 (Hydrochloride) CAS:371935-79-4 PI-103 Hydrochloride is a dual PI3K and mTOR inhibitor with IC 50 s of 8 nM, 88 nM, 48 nM, 150 nM, 20 nM, and 83 nM for p110α , p110β , p110δ , p110γ , mTORC1 , and mTORC2 . PI-103 also inhibits DNA-PK with an IC50 of 2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.