Learn More
Medchemexpress LLC PI-103 hydrochloride | 371935-79-4 | MFCD10687102 | 98.0% | 384.82 | C19H16N4O3.HCl | 10 MG
SDP
Catalog No. 5000211243
Encompass_Preferred
PI-103 (hydrochloride) is a research-grade small-molecule inhibitor that targets class I PI3K isoforms, mTOR, and DNA-PK. It is used as a biochemical and cellular tool to study PI3K/mTOR signaling, apoptosis, and autophagy in cancer and related research.
  • Dual PI3K and mTOR inhibitory activity with low-nanomolar potency.
  • Reported IC50 values: p110α 8 nM; p110β 88 nM; p110δ 48 nM; p110γ 150 nM; mTORC1 20 nM; mTORC2 83 nM; DNA-PK 2 nM.
  • Available in small-mass packs suitable for analytical and research use.
  • Soluble in DMSO and compatible with cell-based and biochemical assays.
  • Used to probe PI3K/mTOR pathway, autophagy, and apoptosis mechanisms in oncology research.
Catalog No. 50-002-11243 Supplier Medchemexpress LLC Supplier No. HY1011510MG
PI-103 (hydrochloride) is a research-grade small-molecule inhibitor that targets class I PI3K isoforms, mTOR, and DNA-PK. It is used as a biochemical and cellular tool to study PI3K/mTOR signaling, apoptosis, and autophagy in cancer and related research.
  • Dual PI3K and mTOR inhibitory activity with low-nanomolar potency.
  • Reported IC50 values: p110α 8 nM; p110β 88 nM; p110δ 48 nM; p110γ 150 nM; mTORC1 20 nM; mTORC2 83 nM; DNA-PK 2 nM.
  • Available in small-mass packs suitable for analytical and research use.
  • Soluble in DMSO and compatible with cell-based and biochemical assays.
  • Used to probe PI3K/mTOR pathway, autophagy, and apoptosis mechanisms in oncology research.