Learn More
Medchemexpress LLC PI-103 hydrochloride | 371935-79-4 | MFCD10687102 | 98.6% | 384.82 g/mol | C19H17ClN4O3 · HCl | 50 MG
SDP
Catalog No. 5000213389
Encompass_Preferred
PI-103 hydrochloride is a research-grade small molecule inhibitor of phosphatidylinositol 3-kinase (PI3K) and mechanistic target of rapamycin (mTOR). It is used in biochemical and cellular studies to probe PI3K/mTOR signaling and DNA-PK activity, and is supplied as a hydrochloride salt for consistent handling in laboratory assays.
  • Dual PI3K and mTOR inhibitor useful for pathway interrogation.
  • Potent biochemical activity with low-nanomolar IC50s against key kinases.
  • Also inhibits DNA-PK, supporting DNA damage response studies.
  • High purity suitable for research applications.
  • Available in multiple sizes for screening or scale-up experiments.
Catalog No. 50-002-13389 Supplier Medchemexpress LLC Supplier No. HY10115A50MG
PI-103 hydrochloride is a research-grade small molecule inhibitor of phosphatidylinositol 3-kinase (PI3K) and mechanistic target of rapamycin (mTOR). It is used in biochemical and cellular studies to probe PI3K/mTOR signaling and DNA-PK activity, and is supplied as a hydrochloride salt for consistent handling in laboratory assays.
  • Dual PI3K and mTOR inhibitor useful for pathway interrogation.
  • Potent biochemical activity with low-nanomolar IC50s against key kinases.
  • Also inhibits DNA-PK, supporting DNA damage response studies.
  • High purity suitable for research applications.
  • Available in multiple sizes for screening or scale-up experiments.