Learn More
Medchemexpress LLC HY-10115A 100mg , PI-103 (Hydrochloride) CAS:371935-79-4 Purity:98%
SDP
Catalog No. 501493251
Encompass_Preferred
Medchemexpress, HY-10115A 100mg PI-103 (Hydrochloride) CAS:371935-79-4 Formula:C19H17ClN4O3 IC50: 8 nM (p110α), 88 nM (p110β), 48 nM (p110δ), 150 nM (p110γ), 2 nM (DNA-PK), 20 nM (mTORC1), 83 nM (mTORC2), 26 nM (PI3KC2β), 850 nM (ATR), 920 nM (ATM), ~1 μM (PI3KC2α), 2.3 μM (hsVPS34), ~50 μM (PI4KIIIβ) Purity:98% PI-103 Hydrochloride is a potent PI3K and mTOR inhibitor with IC 50 s of 8 nM, 88 nM, 48 nM, 150 nM, 20 nM, and 83 nM for p110α , p110β , p110δ , p110γ , mTORC1 , and mTORC2 . PI-103 also inhibits DNA-PK with an IC50 of 2 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3251 Supplier Medchemexpress LLC Supplier No. HY10115A100MG
Medchemexpress, HY-10115A 100mg PI-103 (Hydrochloride) CAS:371935-79-4 Formula:C19H17ClN4O3 IC50: 8 nM (p110α), 88 nM (p110β), 48 nM (p110δ), 150 nM (p110γ), 2 nM (DNA-PK), 20 nM (mTORC1), 83 nM (mTORC2), 26 nM (PI3KC2β), 850 nM (ATR), 920 nM (ATM), ~1 μM (PI3KC2α), 2.3 μM (hsVPS34), ~50 μM (PI4KIIIβ) Purity:98% PI-103 Hydrochloride is a potent PI3K and mTOR inhibitor with IC 50 s of 8 nM, 88 nM, 48 nM, 150 nM, 20 nM, and 83 nM for p110α , p110β , p110δ , p110γ , mTORC1 , and mTORC2 . PI-103 also inhibits DNA-PK with an IC50 of 2 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.