Learn More
Cayman Chemical PI-103 (hydrochloride), Purity: ≥98%, CAS: 371935-79-4, 10 mg
SDP
Catalog No. 502982831
Encompass_Preferred
A potent, cell-permeable, ATP-competitive inhibitor of PI3K family members (IC50 = 2, 8, 20, 26, 48, 83, 88, and 150 nM for DNA-PK, p110α, mTORC1, PI3-KC2β, p110δ, mTORC2, p110β, and p110γ, respectively)
Catalog No. 50-298-2831 Supplier Cayman Chemical Supplier No. 2118710 MG
A potent, cell-permeable, ATP-competitive inhibitor of PI3K family members (IC50 = 2, 8, 20, 26, 48, 83, 88, and 150 nM for DNA-PK, p110α, mTORC1, PI3-KC2β, p110δ, mTORC2, p110β, and p110γ, respectively)