Learn More
Avanti Polar Lipids 1,2-distearoyl-sn-glycero-3-phosphoinositol (ammonium salt), 849412-67-5, 100 ug
Catalog No. NC1018004
Encompass
Molecular Formula: C45H90NO13P, Formula Weight: 884.169, Purity >99%. Inositol phospholipid species are membrane-bound signaling molecules that have been implicated in almost all aspects of cellular physiology, including cellular growth, metabolism, proliferation, and survival.
Catalog No. NC1018004 Supplier Avanti Polar Lipids Supplier No. 850143P100UG
Molecular Formula: C45H90NO13P, Formula Weight: 884.169, Purity >99%. Inositol phospholipid species are membrane-bound signaling molecules that have been implicated in almost all aspects of cellular physiology, including cellular growth, metabolism, proliferation, and survival.