Learn More
Apexbio Technology LLC PI-1840 1401223-22-0 10mg
SDP
Catalog No. 5000088501
Encompass_Preferred
PI-1840 (CAS 1401223-22-0) is a potent selective and rapidly reversible noncovalent inhibitor of the proteasome chymotrypsin-like (CT-L) activity It exhibits an IC50 of 27 nM against CT-L catalytic function of the proteasome with significantly greater selectivity over other proteolytic activities including PGPH-L and T-L In MDA-MB-468 human breast cancer cells PI-1840 inhibits CT-L activity (IC50 0 37 M) suppresses cell viability and proliferation and induces accumulation of CT-L substrates such as I B- p27 and Bax ultimately triggering apoptosis via PARP cleavage and caspase-3 activation In vivo PI-1840 markedly reduces tumor growth in a breast cancer xenograft model This compound is valuable for investigating proteasome function apoptosis and cancer biology
Catalog No. 50-000-88501 Supplier Apexbio Technology LLC Supplier No. B475510
PI-1840 (CAS 1401223-22-0) is a potent selective and rapidly reversible noncovalent inhibitor of the proteasome chymotrypsin-like (CT-L) activity It exhibits an IC50 of 27 nM against CT-L catalytic function of the proteasome with significantly greater selectivity over other proteolytic activities including PGPH-L and T-L In MDA-MB-468 human breast cancer cells PI-1840 inhibits CT-L activity (IC50 0 37 M) suppresses cell viability and proliferation and induces accumulation of CT-L substrates such as I B- p27 and Bax ultimately triggering apoptosis via PARP cleavage and caspase-3 activation In vivo PI-1840 markedly reduces tumor growth in a breast cancer xenograft model This compound is valuable for investigating proteasome function apoptosis and cancer biology