Learn More
Medchemexpress LLC PI-1840 | 1401223-22-0 | 98.8% | 394.47 | 25 MG
SDP
Catalog No. 5000438170
Encompass_Preferred
PI-1840 is a potent and selective chymotrypsin-like (CT-L) inhibitor with an IC50 value of 27 nM. It inhibits cell proliferation and arrests the cell cycle at the G2/M phase. PI-1840 also induces apoptosis and autophagy, and causes the accumulation of proteasome substrates p27, Bax, and IκB-α. It is for research use only.
  • Potent and selective chymotrypsin-like (CT-L) inhibitor
  • IC50 value of 27 nM
  • Inhibits cell proliferation
  • Arrests cell cycle at G2/M phase
  • Induces apoptosis
  • Induces autophagy
  • Induces the accumulation of proteasome substrates p27, Bax, and IκB-α
Catalog No. 50-004-38170 Supplier Medchemexpress LLC Supplier No. 25MGHY12286
PI-1840 is a potent and selective chymotrypsin-like (CT-L) inhibitor with an IC50 value of 27 nM. It inhibits cell proliferation and arrests the cell cycle at the G2/M phase. PI-1840 also induces apoptosis and autophagy, and causes the accumulation of proteasome substrates p27, Bax, and IκB-α. It is for research use only.
  • Potent and selective chymotrypsin-like (CT-L) inhibitor
  • IC50 value of 27 nM
  • Inhibits cell proliferation
  • Arrests cell cycle at G2/M phase
  • Induces apoptosis
  • Induces autophagy
  • Induces the accumulation of proteasome substrates p27, Bax, and IκB-α