Learn More
Selleck Chemical LLC PI-1840
SDP
Catalog No. 5000003566
Encompass_Preferred
PI-1840 is a reversible and selective chymotrypsin-like (CT-L) inhibitor with IC50 of 27 nM with little effects on the other two major proteasome proteolytic activities trypsin-like (T-L) and postglutamyl-peptide-hydrolysis-like (PGPH-L)
Catalog No. 50-000-03566 Supplier Selleck Chemical LLC Supplier No. S746250MG
PI-1840 is a reversible and selective chymotrypsin-like (CT-L) inhibitor with IC50 of 27 nM with little effects on the other two major proteasome proteolytic activities trypsin-like (T-L) and postglutamyl-peptide-hydrolysis-like (PGPH-L)