Learn More
TARGETMOL CHEMICALS INC PI-273 10MG
SDP
Catalog No. 502878390
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. PI-273, a Substrate-Competitive, Specific Small-Molecule Inhibitor of PI4KIIalpha, Inhibits the Growth of Breast Cancer Cells Purity 98.04%
Catalog No. 50-287-8390 Supplier TARGETMOL CHEMICALS INC Supplier No. T1245410MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. PI-273, a Substrate-Competitive, Specific Small-Molecule Inhibitor of PI4KIIalpha, Inhibits the Growth of Breast Cancer Cells Purity 98.04%