Learn More
Medchemexpress LLC HY-103489 1mg Medchemexpress, PI-273 CAS:925069-34-7 Purity:>98%
SDP
Catalog No. 501968123
Encompass_Preferred
Medchemexpress, HY-103489 1mg PI-273 CAS:925069-34-7 PI-273 is a first reversibly and specific phosphatidylinositol 4-kinase (PI4KIIα) inhibitor with an IC50 of 0.47 μM. PI-273 can inhibit breast cancer cell proliferation, block the cell cycle and induce cell apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8123 Supplier Medchemexpress LLC Supplier No. HY1034891MG
Medchemexpress, HY-103489 1mg PI-273 CAS:925069-34-7 PI-273 is a first reversibly and specific phosphatidylinositol 4-kinase (PI4KIIα) inhibitor with an IC50 of 0.47 μM. PI-273 can inhibit breast cancer cell proliferation, block the cell cycle and induce cell apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.