Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pi-273 | 925069-34-7 | MFCD08719772 | 98.0% | 381.90 g·mol^-1 | C16H16ClN3O2S2 | 1 ML
SDP

Catalog No. 5000245355
Encompass_Preferred

A small-molecule inhibitor of phosphatidylinositol 4-kinase IIα supplied as a 10 mM solution in DMSO for research use. The compound has CAS number 925069-34-7, molecular formula C16H16ClN3O2S2, and molecular weight 381.90 g·mol⁻1. Supplied as a 1 mL vial at approximately 98% purity.

  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • Reported subtype-specific inhibitor of PI4KIIα with IC50 ≈0.47 μM.
  • High purity (≈98% by HPLC).
  • Molecular formula C16H16ClN3O2S2; molecular weight 381.90 g·mol⁻1.
  • Soluble in DMSO; suitable for in vitro research applications.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-45355 Supplier Medchemexpress LLC Supplier No. HY10348910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A small-molecule inhibitor of phosphatidylinositol 4-kinase IIα supplied as a 10 mM solution in DMSO for research use. The compound has CAS number 925069-34-7, molecular formula C16H16ClN3O2S2, and molecular weight 381.90 g·mol⁻1. Supplied as a 1 mL vial at approximately 98% purity.

  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • Reported subtype-specific inhibitor of PI4KIIα with IC50 ≈0.47 μM.
  • High purity (≈98% by HPLC).
  • Molecular formula C16H16ClN3O2S2; molecular weight 381.90 g·mol⁻1.
  • Soluble in DMSO; suitable for in vitro research applications.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.