Learn More
Echelon Biosciences Research Labs PI(3,4,5)P3 DiC4 1 mg
SDP
Catalog No. 50720593
Encompass
PI(3,4,5)P3 diC4 (Phosphatidylinositol 3,4,5-trisphosphate diC4) is a synthetic, purified dibutanoyl PI(3,4,5)P3.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,4,5-trisphosphate (PIP3), formed from PI(4,5)P2 though phosphorylation by PI 3-kinase, activates numerous signaling pathways resulting in cell proliferation, growth, survival, glucose transport and protein synthesis. High PIP3 levels from disregulation of PI3-K have been demonstrated in cancer and inflammatory diseases. PIP3 is hydrolyzed by the phosphatases PTEN to PI(4,5)P2 and SHIP to PI(3,4)P2. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-593 Supplier Echelon Biosciences Research Labs Supplier No. P39041MG
PI(3,4,5)P3 diC4 (Phosphatidylinositol 3,4,5-trisphosphate diC4) is a synthetic, purified dibutanoyl PI(3,4,5)P3.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,4,5-trisphosphate (PIP3), formed from PI(4,5)P2 though phosphorylation by PI 3-kinase, activates numerous signaling pathways resulting in cell proliferation, growth, survival, glucose transport and protein synthesis. High PIP3 levels from disregulation of PI3-K have been demonstrated in cancer and inflammatory diseases. PIP3 is hydrolyzed by the phosphatases PTEN to PI(4,5)P2 and SHIP to PI(3,4)P2. This item is non returnable