Learn More
Echelon Biosciences Research Labs PI(3,4)P2 DiC8 1 mg
SDP
Catalog No. 50720572
Encompass
PI(3,4)P2 diC8 (Phosphatidylinositol 3,4-bisphosphate diC8) is a synthetic, purified dioctanoyl PI(3,4)P2.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) is produced in stimulated cells by the action of phosphatidylinositol 3-kinases (PI3Ks) and/or lipid phosphatases (SHIP, TPTE, etc.). PI(3,4)P2 binds the pleckstrin homology (PH)-domain of AKT/PKB leading to cell survival and it also has an important role in podosome formation near focal adhesions. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-572 Supplier Echelon Biosciences Research Labs Supplier No. P34081MG
Add to Cart
Add to Cart
PI(3,4)P2 diC8 (Phosphatidylinositol 3,4-bisphosphate diC8) is a synthetic, purified dioctanoyl PI(3,4)P2.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) is produced in stimulated cells by the action of phosphatidylinositol 3-kinases (PI3Ks) and/or lipid phosphatases (SHIP, TPTE, etc.). PI(3,4)P2 binds the pleckstrin homology (PH)-domain of AKT/PKB leading to cell survival and it also has an important role in podosome formation near focal adhesions. This item is non returnable