Learn More
Echelon Biosciences Research Labs PI(3 5)P2 DiC16 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs P3516100UG
Encompass
PI(3,5)P2 diC16 9Phosphatidylinositol 3,5-bisphosphate diC16) is a synthetic, purified dipalmitoyl PI(3,5)P2.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,5-bisphosphate is in very low abundance and is required for retrograde membrane trafficking from lysosomal and late endosomal compartments to the Golgi and is involved in autophagy. PI(3,5)P2 levels are controlled by the 5-kinase PIKfyve and the 5-phosphatase Fig4/Sac3 and disregulation is linked to several human neuropathies (eg Chacot-Marie-Tooth disease). This item is non returnable
Catalog No. 50-720-588