Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Echelon Biosciences Research Labs AS604850 (PI3-K gamma Inhibitor 2) 5 mg
SDP

Supplier:  Echelon Biosciences Research Labs B03025MG

Encompass

5-(2,2-Difluoro-benzo[1,3]dioxol-5-ylmethylene)-thiazolidine-2,4-dione (AS604850) is an inhibitor of PI 3-K with an IC50 = 250 nM. AS604850 is selective for the -isoform (IC50: PI 3-K = 4.5 M, PI 3-K, PI 3-K > 20 M) and shows no notable activity against a wide array of protein kinases at 1 M.nn


nn

Solubility

nWater: insoluble, DMSO: 5 mg/mLn

Storage

nStore dry between 2-8oC. Stock solutions should be stored frozen (-20oC).n

References

n1) M. Camps et al. "Blockade of PI3Kg suppresses joint inflammation and damage in mouse models of rheumatoid arthritis" Nature Medicine, 2005, 11, 936-943.nn
nn

Documents

nTechnical Data Sheet This item is non returnable

Catalog No. 50-720-015


May include imposed supplier surcharges.
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.