Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Echelon Biosciences Research Labs AS605240 (PI3-K gamma Inhibitor 1) 1 mg
SDP

Catalog No. 50720012
Encompass

5-quinoxilin-6-methylene-1,3-thiazolidine-2,4-dione (AS605240) is an ATP-competitive inhibitor of PI3-K (IC50 = 8 nM). It is selective for the -isoform (IC50: PI 3-K = 60 nM, PI 3-K = 270 nM, PI 3-K = 300 nM) and shows no notable activity against a wide array of protein kinases at 1 M. In addition, AS605240 suppresses the progression of joint inflammation in a mouse model of rheumatoid arthritis and suppresses inflammation in a mouse model of lupus.nn


nn

Solubility

nwater: insoluble, DMSO: 2.5 mg/mLn

Storage

nstore dry between 2-8oC. Stock solutions should be stored frozen (-20oC).n

References

n1) M. Camps et al. "Blockade of PI3Kg suppresses joint inflammation and damage in mouse models of rheumatoid arthritis" Nature Medicine, 2005, 11, 936-943. This item is non returnable

Catalog No. 50-720-012 Supplier Echelon Biosciences Research Labs Supplier No. B03011MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-quinoxilin-6-methylene-1,3-thiazolidine-2,4-dione (AS605240) is an ATP-competitive inhibitor of PI3-K (IC50 = 8 nM). It is selective for the -isoform (IC50: PI 3-K = 60 nM, PI 3-K = 270 nM, PI 3-K = 300 nM) and shows no notable activity against a wide array of protein kinases at 1 M. In addition, AS605240 suppresses the progression of joint inflammation in a mouse model of rheumatoid arthritis and suppresses inflammation in a mouse model of lupus.nn


nn

Solubility

nwater: insoluble, DMSO: 2.5 mg/mLn

Storage

nstore dry between 2-8oC. Stock solutions should be stored frozen (-20oC).n

References

n1) M. Camps et al. "Blockade of PI3Kg suppresses joint inflammation and damage in mouse models of rheumatoid arthritis" Nature Medicine, 2005, 11, 936-943. This item is non returnable

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.