Learn More
Sigma Aldrich Fine Chemicals Biosciences PI 3-Kinase HTRF Assay, 1 plate, 385 wells
Catalog No. NC1771586
Encompass
The PI 3-Kinase HTRF Assay is a high-performance assay kit that provides a universal method for testing all Class I PI3-Kinases in a homogeneous format. This pack size has a screening capacity of 1 plate.
Catalog No. NC1771586 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 33016
The PI 3-Kinase HTRF Assay is a high-performance assay kit that provides a universal method for testing all Class I PI3-Kinases in a homogeneous format. This pack size has a screening capacity of 1 plate.