Learn More
Medchemexpress LLC PI 3 Kinase p150 Antibody (YA2823) | 100 UL
SDP
Catalog No. 5000444416
Encompass_Preferred
The PI 3 Kinase p150 Antibody (YA2823) is a rabbit-derived antibody supplied as a liquid. It is intended for laboratory use in the manufacture of substances and is reactive with human, mouse, and rat samples. This antibody is stable under recommended storage conditions and has an observed molecular weight of 150 kDa.
  • Reacts with human, mouse, and rat samples.
  • Supports Western Blot (WB), Immunohistochemistry (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC) applications.
  • Formulated in rabbit IgG in 10mM phosphate buffered saline, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.
  • Stable for 1 year when stored at -20°C; avoid repeated freeze/thaw cycles.
  • Shipped with blue ice to maintain stability.
Catalog No. 50-004-44416 Supplier Medchemexpress LLC Supplier No. HYP83078100UL
The PI 3 Kinase p150 Antibody (YA2823) is a rabbit-derived antibody supplied as a liquid. It is intended for laboratory use in the manufacture of substances and is reactive with human, mouse, and rat samples. This antibody is stable under recommended storage conditions and has an observed molecular weight of 150 kDa.
  • Reacts with human, mouse, and rat samples.
  • Supports Western Blot (WB), Immunohistochemistry (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC) applications.
  • Formulated in rabbit IgG in 10mM phosphate buffered saline, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.
  • Stable for 1 year when stored at -20°C; avoid repeated freeze/thaw cycles.
  • Shipped with blue ice to maintain stability.