Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PI 3 Kinase p150 Antibody (YA2823) | 50 UL
SDP

Catalog No. 5000444108
Encompass_Preferred

This rabbit recombinant monoclonal antibody, PI 3 Kinase p150 Antibody (YA2823), is designed for laboratory research applications. It is reactive with human, mouse, and rat samples and is supplied in a phosphate-buffered saline solution containing BSA, sodium azide, and glycerol.

  • Rabbit host
  • Reactivity across human, mouse, and rat
  • Suitable for Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC) applications
  • Supplied in 10mM phosphate buffered saline (pH 7.4) with 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide, and 50% glycerol
  • Recommended storage at -20°C for up to 1 year
  • Observed molecular weight of 150 kDa
  • Antibody type for research use

Catalog No. 50-004-44108 Supplier Medchemexpress LLC Supplier No. HYP8307850UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This rabbit recombinant monoclonal antibody, PI 3 Kinase p150 Antibody (YA2823), is designed for laboratory research applications. It is reactive with human, mouse, and rat samples and is supplied in a phosphate-buffered saline solution containing BSA, sodium azide, and glycerol.

  • Rabbit host
  • Reactivity across human, mouse, and rat
  • Suitable for Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC) applications
  • Supplied in 10mM phosphate buffered saline (pH 7.4) with 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide, and 50% glycerol
  • Recommended storage at -20°C for up to 1 year
  • Observed molecular weight of 150 kDa
  • Antibody type for research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.