Learn More
Echelon Biosciences Research Labs PI(3)P DiC4 1 mg
SDP
Supplier:  Echelon Biosciences Research Labs P30041MG
Encompass
PI(3)P diC4 (Phosphatidylinositol 3-phosphate diC4) is a synthetic, purified dibutanoyl PI(3)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3-phosphate is enriched in early endosomes having roles in endosome fusion and receptor sorting and internalization in multivesicular bodies. PI(3)P has also been found at the plasma membrane and is involved in the translocation of the glucose transport protein GLUT4. This item is non returnable
Catalog No. 50-720-559