Learn More
Echelon Biosciences Research Labs PI(3)P DIC8 AMMONIUM SALT 1 mg
SDP
Catalog No. 50720565
Encompass
PI(3)P diC8 (Phosphatidylinositol 3-phosphate diC8) is a synthetic, purified dioctanoyl PI(3)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3-phosphate is enriched in early endosomes having roles in endosome fusion and receptor sorting and internalization in multivesicular bodies. PI(3)P has also been found at the plasma membrane and is involved in the translocation of the glucose transport protein GLUT4. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-565 Supplier Echelon Biosciences Research Labs Supplier No. P3008A1MG
PI(3)P diC8 (Phosphatidylinositol 3-phosphate diC8) is a synthetic, purified dioctanoyl PI(3)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3-phosphate is enriched in early endosomes having roles in endosome fusion and receptor sorting and internalization in multivesicular bodies. PI(3)P has also been found at the plasma membrane and is involved in the translocation of the glucose transport protein GLUT4. This item is non returnable