Learn More
Echelon Biosciences Research Labs PI(3)P GRIP 2.5 ug
SDP
Supplier:  Echelon Biosciences Research Labs G03022.5UG
Encompass
Each vial of PI(3)P Grip contains lyophilized, recombinant GST-tagged p40PX domain. This PI(3)P-specific binding protein can be used as a control reagent with PIP Strips, PIP Arrays, and PIP Plates. It is easily reconstituted in water or buffered solutions. Each 2.5 g vial contains enough protein for use on one PIP Strip or PIP Array. PI(3)P Grip can be detected with commercial anti-GST antibodies This item is non returnable
Catalog No. 50-720-346