Learn More
Echelon Biosciences Research Labs PI(3)P PolyPIPosomes 0.25 mL
SDP
Catalog No. 50720818
Encompass
PolyPIPosomes are polymerized liposomes that show markedly increased stability (up to 6 months) vs. conventional liposomes. A biotin tag is incorporated for easy detection of PIPn-binding proteins with streptavidin reagents. PolyPIPosomes are provided as a 1mM total lipid solution containing 5% PIP. Each order of 250 L or 1 mL of PolyPIPosomes includes 100 L of Control PolyPIPosomes. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-818 Supplier Echelon Biosciences Research Labs Supplier No. YP003250UL
PolyPIPosomes are polymerized liposomes that show markedly increased stability (up to 6 months) vs. conventional liposomes. A biotin tag is incorporated for easy detection of PIPn-binding proteins with streptavidin reagents. PolyPIPosomes are provided as a 1mM total lipid solution containing 5% PIP. Each order of 250 L or 1 mL of PolyPIPosomes includes 100 L of Control PolyPIPosomes. This item is non returnable