Learn More
Selleck Chemical LLC PI-3065
SDP
Catalog No. 5000000239
Encompass_Preferred
PI-3065 is a selective p110 inhibitor with IC50 of 15 nM 70-fold selectivity over other PI3K family members
Catalog No. 50-000-00239 Supplier Selleck Chemical LLC Supplier No. S76235MG
PI-3065 is a selective p110 inhibitor with IC50 of 15 nM 70-fold selectivity over other PI3K family members