Learn More
Apexbio Technology LLC PI-3065, 5mg. Cas: 955977-50-1 MFCD: MFCD28167771. P110δkinase inhibitor
SDP
Supplier:  Apexbio Technology LLC B46605
Encompass_Preferred
PI-3065 is a small molecule and selective inhibitor of p110kinase with an Ki value of 1.5nM and IC50 value of 5nM [1].PI-3065 has been reported to selectively inhibit p110kinase with Ki values of 15nM, 110nM, 130nM and 940nM Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5354