Learn More
Echelon Biosciences Research Labs PI(4,5)P2 PIP Beads 1mL
SDP
Catalog No. 50720689
Encompass
PI(4,5)P2 PIP BeadsTM are composed of agarose with 10 nanomoles of bound PI(4,5)P2 per mL of beads, enough for several protein binding experiments. Each 1 mL order of PIP Beads includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-689 Supplier Echelon Biosciences Research Labs Supplier No. PB045A1ML
PI(4,5)P2 PIP BeadsTM are composed of agarose with 10 nanomoles of bound PI(4,5)P2 per mL of beads, enough for several protein binding experiments. Each 1 mL order of PIP Beads includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable